Mevastatin 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (1S,7S,8S,8aR)-8-(2-((2R,4R)-4-Hydroxy-6-oxotetrahydro-2H-pyran-2-yl)ethyl)-7-methyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl (S)-2-methylbutanoate, Compactin

J61357-LB0 J61357-MA J61357-MB
CAAAJ61357-LB0EA 128.28 CAD
CAAAJ61357-LB0 CAAAJ61357-MA CAAAJ61357-MB
Mevastatin 98%
Mevastatin

Inhibitor of HMG-CoA reductase. Induces apoptosis


Inhibitor of HMG-CoA reductase. Induces apoptosis


Inhibitor of HMG-CoA reductase. Induces apoptosis

Formula: C₂₃H₃₄O₅
MW: 390.52 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05662341
CAS Number: 73573-88-3
Merck Index: 14,6164

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR