5-Iodo-2'-deoxycytidine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J61356-03
J61356-04
CAAAJ61356-03EA
554.93
CAD
CAAAJ61356-03
CAAAJ61356-04
5-Iodo-2'-deoxycytidine 99%
5-Iodo-2'-deoxycytidine
Powder
Purity: 99%
CAS No.: 611-53-0
Molecular Formula: C9H12IN3O4
Formula Weight: 353.11
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Melting Point: 180-183°
MDL No.: MFCD00038063
Formula:
C₉H₁₂IN₃O₄ Melting Pt: 180…183 °C Storage Temperature: Freezer |
MDL Number:
MFCD00038063 CAS Number: 611-53-0 EINECS: 210-269-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...