Azure A
Supplier: Thermo Scientific Chemicals
|
Warning
|
J61346-14
J61346-22
CAAAJ61346-14EA
64.76
CAD
CAAAJ61346-14
CAAAJ61346-22
Azure A
Azure A
For staining cell granules and tissues.
Powder
For staining cell granules and tissues
Formula:
C₁₄H₁₄ClN₃S Melting Pt: 290 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012112 CAS Number: 531-53-3 EINECS: 208-510-7 Merck Index: 14,927 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...