Phosphate, 0.2 M buffer soln., pH 7.5, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J61324-AP J61324-K2
CAAAJ61324-APEA 75.68 CAD
CAAAJ61324-AP CAAAJ61324-K2
Phosphate, 0.2 M buffer soln., pH 7.5, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR