Streptomycin sulfate cell culture reagent

Supplier: Thermo Scientific Chemicals
Danger

J61299-18 J61299-22
CAAAJ61299-18EA 169.69 CAD
CAAAJ61299-18 CAAAJ61299-22
Streptomycin sulfate cell culture reagent
Streptomycin sulphate

Inhibits initiation and causes misreading of rRNA in protein synthesis.


Powder


Inhibits initiation and causes misreading of rRNA in protein synthesis

Formula: C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
Melting Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00037023
CAS Number: 3810-74-0
EINECS: 223-286-0
ADR: 6.1,III
Merck Index: 14,8826

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR