Sodium acetate, 3 M aq. soln., pH 4.5, autoclaved, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J61288-AK J61288-EQE
CAAAJ61288-AKEA 225.1 CAD
CAAAJ61288-AK CAAAJ61288-EQE
Sodium acetate, 3 M aq. soln., pH 4.5, autoclaved, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 127-09-3
Molecular Formula: C2H3NaO2
Formula Weight: 82.03
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00012459

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR