Nimodipine ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Isopropyl 5-(2-methoxyethyl) 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, Isopropyl 2-Methoxyethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate

J61287-03 J61287-ME
CAAAJ61287-03EA 596.92 CAD
CAAAJ61287-03 CAAAJ61287-06 CAAAJ61287-ME
Nimodipine ≥98%
Nimodipine

Potent L-type calcium channel antagonist


Powder


Applications: Potent L-type calcium channel antagonist

Formula: C₂₁H₂₆N₂O₇
MW: 418.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153848
CAS Number: 66085-59-4
EINECS: 266-127-0
Merck Index: 14,6551

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR