(+/-)-Octopamine hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: alpha-(Aminomethyl)-4-hydroxybenzyl alcohol hydrochloride, 4-(2-Amino-1-hydroxyethyl)phenol hydrochloride, dl-Octopamine hydrochloride, 4-(2-amino-1-hydroxyethyl)phenol hydrochloride, Octopamine hydrochloride

J61281-09 J61281-03 J61281-06
CAAAJ61281-09EA 311.99 CAD
CAAAJ61281-09 CAAAJ61281-03 CAAAJ61281-06
(+/-)-Octopamine hydrochloride 99%
(+/-)-Octopamine hydrochloride

alpha-adrenoceptor agonist; invertebrate neurotransmitter and neuromodulator


Powder


Applications: alpha-adrenoceptor agonist; invertebrate neurotransmitter and neuromodulator

Formula: C₈H₁₂ClNO₂
MW: 189.639 g/mol
Melting Pt: 170 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012881
CAS Number: 770-05-8
EINECS: 212-216-4
Merck Index: 14,6759

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR