Puromycin dihydrochloride 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J61278-MB
J61278-MC
J61278-ME
CAAAJ61278-MBEA
149.44
CAD
CAAAJ61278-MB
CAAAJ61278-MC
CAAAJ61278-ME
Puromycin dihydrochloride 99%
Puromycin dihydrochloride
Causes premature chain termination in protein synthesis
Powder
Causes premature chain termination in protein synthesis
Formula:
C₂₂H₂₉N₇O₅·2HCl Melting Pt: 168…170 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00012691 CAS Number: 58-58-2 EINECS: 200-387-8 Merck Index: 14,7943 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...