Polyethyleneimine, M.W. 60,000 50% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Polyaziridine, Corcat, Polyethylenimine, Polymin P, Polyaziridine Mw 25000, Aziridine, PEI, Polyethylenimine (branched), Ethyleneimine Polymer, Poly(ethyleneimine)

J61270-22 J61270-30
CAAAJ61270-22EA 150.45 CAD
CAAAJ61270-22 CAAAJ61270-30
Polyethyleneimine, M.W. 60,000 50% (w/w) in aqueous solution
Polyethyleneimine

A protein precipitant used to purify proteins


Liquid


Applications: A protein precipitant used to purify proteins

Formula: (CH2CH2NH)n
Density: 1.07
Storage Temperature: Ambient
MDL Number: MFCD00084427
CAS Number: 9002-98-6
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR