U0126 ≥99%

Supplier: Thermo Scientific Chemicals
Warning

J61246-MB J61246-MC
CAAAJ61246-MBEA 525.89 CAD
CAAAJ61246-MB CAAAJ61246-MC CAAAJ61246-EXH
U0126 ≥99%
U0126

Selective inhibitor of mitogen-activated protein kinases MEK-1 and MEK-2


Powder


Soluble in DMSO. Poorly soluble in ethanol and water

Formula: C₁₈H₁₆N₆S₂
MW: 380.49 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01861186
CAS Number: 109511-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR