AMPSO, 0.2 M Buffer Solutions, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J61238-AE J61238-AK J62328-AE J62328-AK J63354-AK
CAAAJ61238-AEEA 170.01 CAD
CAAAJ61238-AE CAAAJ61238-AK CAAAJ61467-AE CAAAJ61467-AK CAAAJ62328-AE CAAAJ62328-AK CAAAJ63354-AE CAAAJ63354-AK
AMPSO, 0.2 M Buffer Solutions, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 102029-60-7
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00067500


Caution: Storage Temperature: 2 to 8 °C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR