Dextran, M.W. ~ 20,000

Supplier: Thermo Scientific Chemicals

J61216-09 J61216-18 J61216-22 J61216-36
CAAAJ61216-09EA 91.48 CAD
CAAAJ61216-09 CAAAJ61216-18 CAAAJ61216-22 CAAAJ61216-36
Dextran, M.W. ~ 20,000
Dextran

Powder


CAS No.: 9004-54-0 Molecular Formula: (C6H10O5)n Physical Form: Powder Appearance: White to off-white Merck Reference: 14,2948 MDL No.: MFCD00130935

Formula: (C₆H₁₀O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00130935
CAS Number: 9004-54-0
EINECS: 232-677-5
Merck Index: 14,2948

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR