Ribonucleic acid (from bakers yeast)
Supplier: Thermo Scientific Chemicals
J61215-03
J61215-MC
J61215-ME
CAAAJ61215-03EA
1681.09
CAD
CAAAJ61215-03
CAAAJ61215-MC
CAAAJ61215-ME
Ribonucleic acid (from bakers yeast)
Ribonucleic acid
Ribonucleic acid (RNA) from baker's yeast S. cerevisiae may be used as a substrate for studying ribonuclease activities of enzymes such as RNAase(s). It is also suitable for use as a carrier in nucleic acid purification and precipitation.
- Off-white to pale brown powder
- Soluble in water
- Store at cold temperature
Suitable for use as a ribonuclease substrate.
Caution: Not suitable for use as a carrier in nucleic acid purification and precipitation.
Storage Temperature:
Refrigerator |
MDL Number:
MFCD00132195 CAS Number: 63231-63-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...