Fluoxetine hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Danger

J61197-MD J61197-MF
CAAAJ61197-MDEA 780.52 CAD
CAAAJ61197-MD CAAAJ61197-MF
Fluoxetine hydrochloride 99%
Fluoxetine hydrochloride

A selective serotonin reuptake inhibitor


Powder


Applications: A selective serotonin reuptake inhibitor

Formula: C₁₇H₁₈F₃NO·HCl
Melting Pt: 158…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00214288
CAS Number: 56296-78-7
EINECS: 260-101-2
Merck Index: 14,4185

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR