TAPS sodium salt (3-(Tris(hydroxymethyl)methylamino)propane-1-sulfonic acid sodium salt) 98%

Supplier: Thermo Scientific Chemicals
Warning

J61189-14 J61189-22
CAAAJ61189-14EA 81.51 CAD
CAAAJ61189-14 CAAAJ61189-22
TAPS sodium salt (3-(Tris(hydroxymethyl)methylamino)propane-1-sulfonic acid sodium salt) 98%
TAPS sodium salt (3-(Tris(hydroxymethyl)methylamino)propane-1-sulphonic acid sodium salt)

Zwitterionic buffer


Powder


Zwitterionic buffer

Formula: C₇H₁₆NNaO₆S
Storage Temperature: Ambient
MDL Number: MFCD00069903
CAS Number: 91000-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR