Benzydamine hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

J61180-06 J61180-14
CAAAJ61180-06EA 128.75 CAD
CAAAJ61180-06 CAAAJ61180-14
Benzydamine hydrochloride
Benzydamine hydrochloride

Nonsteroidal analgesic and anti-inflammatory compound. A prostaglandin synthase inhibitor


Crystalline powder


Applications: Nonsteroidal analgesic and anti-inflammatory compound. A prostaglandin synthase inhibitor

Formula: C₁₉H₂₃N₃O
Melting Pt: 156…161 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00078957
CAS Number: 132-69-4
EINECS: 205-076-0
Merck Index: 14,1122

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR