Maleate, 0.2 M buffer soln., pH 6.5, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J61156-AK J61156-AP
CAAAJ61156-AKEA 175.14 CAD
CAAAJ61156-AK CAAAJ61156-AP
Maleate, 0.2 M buffer soln., pH 6.5, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 676-46-0
Physical Form: Liquid
MDL No.: MFCD00012466

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR