Tyr-Gly-Gly-Phe-Met
Supplier: Thermo Scientific Chemicals
Endogenous opioid peptide that also functions as a growth factor on many cell types at a receptor that is distinct from the neuronal opioid receptors.
- EINECS: 261-335-8
CAS No.: 58569-55-4
Molecular Formula: C27H35N5O7S
Formula Weight: 573.67
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00076415
Endogenous opioid peptide that also functions as a growth factor on many cell types at a receptor that is distinct from the neuronal opioid receptors
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...