5,7-Dichlorokynurenic acid monohydrate

Supplier: Thermo Scientific Chemicals
Warning

J61138-MA J61138-MF
CAAAJ61138-MAEA 248.66 CAD
CAAAJ61138-MA CAAAJ61138-MC CAAAJ61138-MF
5,7-Dichlorokynurenic acid monohydrate
5,7-Dichlorokynurenic acid monohydrate

Solid


CAS No.: 190908-40-8 Molecular Formula: C10H5Cl2NO3.H2O Formula Weight: 276.08 (258.06anhy) Storage Temperature: 2°C to 8°C Physical Form: Solid Appearance: Pale yellow Solubility: Soluble in 1 eq. NaOH to 50 mM MDL No.: MFCD11045951

Formula: C₁₀H₅Cl₂NO₃·H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD11045951
CAS Number: 190908-40-8
EINECS: 633-778-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR