Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate 98%
Supplier: Thermo Scientific Chemicals
J61125-06
J61125-09
J61125-14
CAAAJ61125-06EA
231.28
CAD
CAAAJ61125-06
CAAAJ61125-09
CAAAJ61125-14
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate 98%
Adenosine 5'-triphosphate disodium salt (ATP disodium salt) hydrate
Powder
Purity: 98%
CAS No.: 34369-07-8
Molecular Formula: C10H14N5Na2O13P3
Formula Weight: 551.15
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Sensitivity: Light Sensitive and Hygroscopic
Appearance: White
Merck Reference: 14,155
MDL No.: MFCD00080339
Formula:
C₁₀H₁₄N₅Na₂O₁₃P₃·xH₂O Storage Temperature: Freezer |
MDL Number:
MFCD00150755 CAS Number: 34369-07-8 EINECS: 213-579-1 Merck Index: 14,155 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...