Agarose, high melting , medium resolution for DNA separations

Supplier: Thermo Scientific Chemicals

J61123-14 J61123-22 J61123-36
CAAAJ61123-14EA 252.03 CAD
CAAAJ61123-14 CAAAJ61123-22 CAAAJ61123-36
Agarose, high melting , medium resolution for DNA separations
Agarose

For standard DNA applications


It is involved in the purification of protein by using column-based preparative scale separation as in gel filtration chromatography and ion exchange chromatography.


Caution: Moisture sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong acids.

Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR