Elastatinal
Supplier: Thermo Scientific Chemicals
J61120-LB0
J61120-MB
CAAAJ61120-LB0EA
379.78
CAD
CAAAJ61120-LB0
CAAAJ61120-MB
Elastatinal
Elastatinal
Irreversible elastase inhibitor
CAS No.: 51798-45-9
Molecular Formula: C21H36N8O7
Formula Weight: 512.60
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to pale yellow
Beilstein Reference: 873543
MDL No.: MFCD00151180
Irreversible elastase inhibitor
Formula:
C₂₁H₃₆N₈O₇ Storage Temperature: Freezer |
MDL Number:
MFCD00151180 CAS Number: 51798-45-9 EINECS: 257-426-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...