Succinate, 0.2 M Buffer Soln., pH 5.5, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J61100-AK J61100-AP
CAAAJ61100-AKEA 170.01 CAD
CAAAJ61100-AK CAAAJ61100-AP
Succinate, 0.2 M Buffer Soln., pH 5.5, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 150-90-3
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00002789

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR