Histamine diphosphate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
J61088-06
J61088-14
CAAAJ61088-06EA
143.97
CAD
CAAAJ61088-06
CAAAJ61088-14
Histamine diphosphate 98%
Histamine diphosphate
Endogenous H1 and H2 agonist
Powder
Applications: Endogenous H1 and H2 agonist
Formula:
C₅H₉N₃·2H₃PO₄ Melting Pt: 128…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064316 CAS Number: 51-74-1 EINECS: 200-118-4 Merck Index: 14,4719 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...