Phosphate, 0.5 M buffer soln., pH 8.5, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J61086-AK J61086-AP
CAAAJ61086-AKEA 100.82 CAD
CAAAJ61086-AK CAAAJ61086-AP
Phosphate, 0.5 M buffer soln., pH 8.5, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR