Tetranitro blue tetrazolium 97%

Supplier: Thermo Scientific Chemicals
Danger

J61080-03 J61080-MC
CAAAJ61080-03EA 1067.78 CAD
CAAAJ61080-03 CAAAJ61080-MC
Tetranitro blue tetrazolium 97%
Tetranitro blue tetrazolium

Redox indicator for enzymes


Powder


Redox indicator for enzymes

Formula: C₄₀H₂₈Cl₂N₁₂O₁₀
Melting Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00036338
CAS Number: 1184-43-6
EINECS: 214-665-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR