Jenner's stain stain for hematology

Supplier: Thermo Scientific Chemicals

Synonyms: Eosin−Methylene Blue

J61077-14 J61077-22
CAAAJ61077-14EA 59.04 CAD
CAAAJ61077-14 CAAAJ61077-22
Jenner's stain stain for hematology
Jenner's stain

For use in staining blood smears


Powder


Applications: For use in staining blood smears

Storage Temperature: Ambient
MDL Number: MFCD00081733
CAS Number: 62851-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR