Castanospermine 99%

Supplier: Thermo Scientific Chemicals
Warning

J61071-MC J61071-ME
CAAAJ61071-MCEA 677.85 CAD
CAAAJ61071-MC CAAAJ61071-ME
Castanospermine 99%
Castanospermine

Potent alpha and beta-glucosidase inhibitor.


Solid


Applications: Potent alpha and beta-glucosidase inhibitor.

Formula: C₈H₁₅NO₄
Melting Pt: 213…217 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00017555
CAS Number: 79831-76-8
Merck Index: 14,1896

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR