Pyronin G
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Pyronin Y, Pyronine Y, 3,6-Bis(dimethylamino)xanthylium chloride
J61068-03
J61068-06
J61068-09
CAAAJ61068-03EA
63.5
CAD
CAAAJ61068-03
CAAAJ61068-06
CAAAJ61068-09
Pyronin G
Pyronine G
Marker dye for RNA useful in acid buffer systems
Crystalline powder
Marker dye for RNA useful in acid buffer systems
Formula:
C₁₇H₁₉ClN₂O Melting Pt: 250…260 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011725 CAS Number: 92-32-0 EINECS: 202-147-8 Merck Index: 14,8007 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...