IGEPAL® CA-630

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Octylphenyl-polyethylene glycol

J61055-AE J61055-AP
CAAAJ61055-AEEA 121.41 CAD
CAAAJ61055-AE CAAAJ61055-AP
IGEPAL® CA-630
IGEPAL® CA-630

Detergent, equivalent to Nonidet P-40. For solubilizing membrane proteins


IGEPAL® CA-630 is used as a detergent in paper industry, hand cleaners, metal cleaners. It is suitable for solubilization, isolation and purification of membrane protein complexes and in two-dimensional gel electrophoresis.


Applications: Detergent, equivalent to Nonidet P-40. For solubilizing membrane proteins


Caution: IGEPAL® is a registered trademark of Rhodia Operations. Incompatible with oxidizing agents and reducing agents.

Formula: (C₂H₄O)nC₁₄H₂₂O
Density: 1.060
Storage Temperature: Ambient
MDL Number: MFCD00132505
CAS Number: 9002-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR