Denatonium benzoate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Benzyl-2-((2,6-dimethylphenyl)amino)-N,N-diethyl-2-oxoethanaminium benzoate

J61048-03 J61048-06 J61048-14
CAAAJ61048-03EA 72.13 CAD
CAAAJ61048-03 CAAAJ61048-06 CAAAJ61048-14
Denatonium benzoate
Denatonium benzoate

The bitterest compound known


Powder


The bitterest compound known

Formula: C₂₈H₃₄N₂O₃
MW: 446.59 g/mol
Melting Pt: 164…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00031578
CAS Number: 3734-33-6
EINECS: 223-095-2
Merck Index: 14,2891

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR