Octyl-β-D-thioglucopyranoside 98+%

Supplier: Thermo Scientific Chemicals

J61028-06 J61028-09 J61028-88
CAAAJ61028-06EA 868.62 CAD
CAAAJ61028-06 CAAAJ61028-09 CAAAJ61028-88
Octyl-β-D-thioglucopyranoside 98+%
Octyl β-D-thioglucopyranoside

Non-ionic detergent used for the solubilization of membrane proteins.


Crystalline powder


Non-ionic detergent used for the solubilization of membrane proteins

Formula: C₁₄H₂₈O₅S
Melting Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00012189
CAS Number: 85618-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR