Dacarbazine

Supplier: Thermo Scientific Chemicals
Danger

J61023-03 J61023-ME
CAAAJ61023-03EA 719.69 CAD
CAAAJ61023-03 CAAAJ61023-ME
Dacarbazine
Dacarbazine

Used in treatment of malignant melanoma and sarcomas. Generates methyl adducts to purine bases in DNA


Powder


Applications: Used in treatment of malignant melanoma and sarcomas. Generates methyl adducts to purine bases in DNA

Formula: C₆H₁₀N₆O
Melting Pt: 210…212 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00057167
CAS Number: 4342-03-4
EINECS: 224-396-1
Merck Index: 14,2798

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR