TRIS, 0.5 M Buffer Solutions, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J61009-AP J61009-K2 J61141-AP J61141-K2 J61144-AP J61144-K2 J61537-K2 J62131-AP J62131-K2 J62287-AP J62287-K2 J62552-AP J62552-K2 J62778-AP J62778-K2 J62944-AP J62944-K2 J63134-K2 J63735-AP J63735-K2
CAAAJ61009-APEA 106.19 CAD
CAAAJ61009-AP CAAAJ61009-K2 CAAAJ61141-AP CAAAJ61141-K2 CAAAJ61144-AP CAAAJ61144-K2 CAAAJ61213-AP CAAAJ61213-K2 CAAAJ61537-AP CAAAJ61537-K2 CAAAJ61787-AP CAAAJ61787-K2 CAAAJ61944-AP CAAAJ61944-K2 CAAAJ62131-AP CAAAJ62131-K2 CAAAJ62287-AP CAAAJ62287-K2 CAAAJ62551-AP CAAAJ62551-K2 CAAAJ62552-AP CAAAJ62552-K2 CAAAJ62736-AP CAAAJ62736-K2 CAAAJ62778-AP CAAAJ62778-K2 CAAAJ62944-AP CAAAJ62944-K2 CAAAJ63134-AP CAAAJ63134-K2 CAAAJ63735-AP CAAAJ63735-K2
TRIS, 0.5 M Buffer Solutions, Thermo Scientific
Buffers Biological Buffers

Tris (hydroxymethyl) aminomethane.


CAS No.: 77-86-1
MDL No.: MFCD00004679

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR