India ink solution 0.2% in PBS
Supplier: Thermo Scientific Chemicals
J61007-AP
J61007-K2
CAAAJ61007-APEA
159.58
CAD
CAAAJ61007-AP
CAAAJ61007-K2
India ink solution 0.2% in PBS
India ink
For negative staining of microorganisms
Liquid
Applications: For negative staining of microorganisms
Storage Temperature:
Ambient |
MDL Number:
MFCD00147244 CAS Number: 8046-52-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...