Vinpocetine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (41S,13aS)-Ethyl 13a-ethyl-2,3,41,5,6,13a-hexahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate

J61000-FPL J61000-MC
CAAAJ61000-FPLEA 278.92 CAD
CAAAJ61000-FPL CAAAJ61000-MC
Vinpocetine 98%
Vinpocetine

Specific calcium-calmodulin-dependent phosphodiesterase inhibitor


Powder


Specific calcium-calmodulin-dependent phosphodiesterase inhibitor

Formula: C₂₂H₂₆N₂O₂
MW: 350.46 g/mol
Melting Pt: 147…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00211233
CAS Number: 42971-09-5
EINECS: 256-028-0
Merck Index: 14,9991

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR