Tyramine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Hydroxyphenethylamine, 4-(2-Aminoethyl)phenol
J60990-03
J60990-14
J60990-06
CAAAJ60990-03EA
37.87
CAD
CAAAJ60990-03
CAAAJ60990-14
CAAAJ60990-06
Tyramine 98+%
Tyramine
Reagent for fluorescence enzyme immunoassay
Powder
Reagent for fluorescence enzyme immunoassay
Formula:
C₈H₁₁NO Boiling Pt: 175…181 °C/8 mm Hg Melting Pt: 160…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008193 CAS Number: 51-67-2 EINECS: 200-115-8 UN: 3263 ADR: "8",III Merck Index: 14,9835 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...