Dextran, M.W. ~ 75,000
Supplier: Thermo Scientific Chemicals
J60989-09
J60989-18
J60989-22
J60989-36
CAAAJ60989-09EA
91.8
CAD
CAAAJ60989-09
CAAAJ60989-18
CAAAJ60989-22
CAAAJ60989-36
Dextran, M.W. ~ 75,000
Dextran
Powder
CAS No.: 9004-54-0
Molecular Formula: (C6H10O5)n
Physical Form: Powder
Appearance: White to off-white
Solubility: Soluble in water, DMF and glycerol
Merck Reference: 14,2948
MDL No.: MFCD00130935
Formula:
(C₆H₁₀O₅)n Storage Temperature: Ambient |
MDL Number:
MFCD00130935 CAS Number: 9004-54-0 EINECS: 232-677-5 Merck Index: 14,2948 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...