Soybeans Trypsin Inhibitor
Supplier: Thermo Scientific Chemicals
J60982-03
J60982-MC
CAAAJ60982-03EA
2213.42
CAD
CAAAJ60982-03
CAAAJ60982-MC
Soybeans Trypsin Inhibitor
Proteins and Peptides
Powder
CAS No.: 9035-81-8
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Off-white to pale brown
Solubility: Dissolve in 1x PBS at 1-10mg/ml for stock solution
MDL No.: MFCD00082103
Applications: Inhibits the pancreatic enzyme trypsin
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...