Diosgenin
Supplier: Thermo Scientific Chemicals
Synonyms:
Nitogenin, (2'R,4S,5'R,6aR,6bS,8aS,8bR,9S,11aS,12aS,12bS)-5',6a,8a,9-Tetramethyl-1,3,3',4,4',5,5',6,6a,6b,6',7,8,8a,8b,9,11a,12,12a,12b-icosahydrospiro[naphtho[2',1':4,5]indeno[2,1-b]furan-10,2'-pyran]-4-ol
J60976-06
J60976-14
J60976-22
CAAAJ60976-06EA
108.07
CAD
CAAAJ60976-06
CAAAJ60976-14
CAAAJ60976-22
Diosgenin
Diosgenin
Useful for synthesis of steroid products. Can be converted to pregnenolone and progesterone
Powder
Applications: Useful for synthesis of steroid products. Can be converted to pregnenolone and progesterone
Formula:
C₂₇H₄₂O₃ Melting Pt: 200…206 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016887 CAS Number: 512-04-9 EINECS: 208-134-3 Merck Index: 14,3295 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...