Adenosine-3',5'-cyclic monophosphate (cAMP) 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3',5'-cAMP, 3′,5′-Cyclic AMP, Adenosine 3′,5′-cyclophosphate, (4aR,6R,7R,7aS)-6-(6-Amino-9H-purin-9-yl)-2,7-dihydroxytetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinine 2-oxide

J60936-03 J60936-06
CAAAJ60936-03EA 313.79 CAD
CAAAJ60936-03 CAAAJ60936-06
Adenosine-3',5'-cyclic monophosphate (cAMP) 98%
Adenosine-3',5'-cyclic monophosphate (cAMP)

Crystalline powder


Purity: 98% CAS No.: 60-92-4 Molecular Formula: C10H12N5O6P Formula Weight: 329.21 Storage Temperature: -30°C to -10°C Physical Form: Crystalline powder Appearance: White to off-white Melting Point: 260° dec. Beilstein Reference: 52645 MDL No.: MFCD00005845

Formula: C₁₀H₁₂N₅O₆P
MW: 329.21 g/mol
Melting Pt: 260 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00005845
CAS Number: 60-92-4
EINECS: 200-492-9
UN: 3261
ADR: "8",III
Merck Index: 14,2708

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR