Agarose Gel Loading Dye 6X for Electrophoresis, No Xylene Cyanol FF, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60933-AC
J60933-AD
CAAAJ60933-ACEA
125.07
CAD
CAAAJ60933-AC
CAAAJ60933-AD
Agarose Gel Loading Dye 6X for Electrophoresis, No Xylene Cyanol FF, Thermo Scientific
Stains
Electrophoresis Stains
6X DNA Loading Buffer is a pre-mixed solution used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels.
- No Xylene cyanol FF
- Contains 0.25% bromophenol blue and 30% glycerol
- Liquid, fully misicible in water
- Store at cold conditions
It is a pre-mixed loading buffer containing two different dyes xylene cyanol FF for visual tracking of DNA migration during agarose gel electrophoresis.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...