MES, 0.2 M buffer soln., pH 6.0, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60930-AK
J60930-AP
CAAAJ60930-AKEA
150.07
CAD
CAAAJ60930-AK
CAAAJ60930-AP
MES, 0.2 M buffer soln., pH 6.0, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 145224-94-8
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00283848
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...