Novobiocin sodium salt ≥95%

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium 4-((7-(((2R,3R,4S,5R)-4-(carbamoyloxy)-3-hydroxy-5-methoxy-6,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-4-hydroxy-8-methyl-2-oxo-2H-chromen-3-yl)carbamoyl)-2-(3-methylbut-2-en-1-yl)phenolate, Novobiocin sodium

J60928-03 J60928-06 J60928-09
CAAAJ60928-03EA 104.88 CAD
CAAAJ60928-03 CAAAJ60928-06 CAAAJ60928-09
Novobiocin sodium salt ≥95%
Novobiocin sodium salt

Topoisomerase II inhibitor


CAS No.: 1476-53-5 Molecular Formula: C31H35N2NaO11 Formula Weight: 634.61 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to pale brown Melting Point: 215° Solubility: Soluble in water at 100mg/ml. Also soluble in ethanol or DMSO Beilstein Reference: 3892910 MDL No.: MFCD00066541


Applications: Topoisomerase II inhibitor

Formula: C₃₁H₃₅N₂NaO₁₁
MW: 634.614 g/mol
Melting Pt: 215 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066541
CAS Number: 1476-53-5
EINECS: 216-023-6
Merck Index: 14,6722

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR