Sodium acetate, 3 M aq. soln., pH 7.0, autoclaved, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60899-AK
J60899-EQE
CAAAJ60899-AKEA
97.49
CAD
CAAAJ60899-AK
CAAAJ60899-EQE
Sodium acetate, 3 M aq. soln., pH 7.0, autoclaved, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 127-09-3
Molecular Formula: C2H3NaO2
Formula Weight: 82.03
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00012459
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...