PIPES, 0.5 M Buffer Solutions, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J61224-AE J61224-AK J61406-AE J61406-AK J61786-AE J61786-AK J62224-AE J63234-AE J63234-AK J63617-AE J63617-AK
CAAAJ60891-AEEA 162.71 CAD
CAAAJ60891-AE CAAAJ60891-AK CAAAJ61224-AE CAAAJ61224-AK CAAAJ61406-AE CAAAJ61406-AK CAAAJ61786-AE CAAAJ61786-AK CAAAJ62224-AE CAAAJ62224-AK CAAAJ63234-AE CAAAJ63234-AK CAAAJ63617-AE CAAAJ63617-AK
PIPES, 0.5 M Buffer Solutions, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 5625-37-6
Appearance: Colorless
MDL No.: MFCD00006159


Caution: Storage Temperature: 2 to 8 °C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR