Nicorandil ≥98%

Supplier: Thermo Scientific Chemicals
Danger

J60879-03 J60879-MD
CAAAJ60879-03EA 803.86 CAD
CAAAJ60879-03 CAAAJ60879-MD
Nicorandil ≥98%
Nicorandil

A vasodilator that activates ATP-sensitive potassium channels


Powder


Applications: A vasodilator that activates ATP-sensitive potassium channels

Formula: C₈H₉N₃O₄
Melting Pt: 92…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00186520
CAS Number: 65141-46-0
EINECS: 265-514-1
Merck Index: 14,6521

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR