TRIS-buffered saline (TBS, 20X) pH 7.4, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60877-K2
J60877-K3
CAAAJ60877-K2EA
116.48
CAD
CAAAJ60877-K2
CAAAJ60877-K3
TRIS-buffered saline (TBS, 20X) pH 7.4, Thermo Scientific
Buffers
Electrophoresis Buffers
Washing Buffers
Liquid
CAS No.: 77-86-1
Physical Form: Liquid
MDL No.: MFCD00132476
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...