Miconazole 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-(2-((2,4-Dichlorobenzyl)oxy)-2-(2,4-dichlorophenyl)ethyl)-1H-imidazole

J60872-03 J60872-06 J60872-14
CAAAJ60872-03EA 89.36 CAD
CAAAJ60872-03 CAAAJ60872-06 CAAAJ60872-14
Miconazole 97%
Miconazole

Inhibits cytochrome P450-dependent 14-alpha-demethylase


Powder


Applications: Inhibits cytochrome P450-dependent 14-alpha-demethylase

Formula: C₁₈H₁₄Cl₄N₂O
MW: 416.123 g/mol
Melting Pt: 83…97 °C
Storage Temperature: Freezer
MDL Number: MFCD00216019
CAS Number: 22916-47-8
EINECS: 245-324-5
Merck Index: 14,6178

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR